Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249285.1 | NP_001236214.1 | 1 - 2 | 125 | 88 | |
GenBank mRNA | AJ276866.1 | - | 2 - 3 | 299 | 88 | |
AY230156.1 | - | 1 - 2 | 125 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |