Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249016.1 | NP_001235945.1 | 6 - 7 | 871 | 79 | |
GenBank mRNA | FJ795036.1 | - | 6 - 7 | 871 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Glycine max Probe spans exons |