Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015555.hg.1 |
TC05001069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015555.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015555.hg.1 |
ENST00000376852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015555.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015556.hg.1 |
TC05001069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015556.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015556.hg.1 |
ENST00000376852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015556.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015554.hg.1 |
TC05001069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015554.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015554.hg.1 |
ENST00000376852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015554.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015553.hg.1 |
TC05001069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015553.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05015553.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168703.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168703.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168703.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168703.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168703.hg.1 |
TC0500009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168705.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168705.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168705.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168705.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168705.hg.1 |
TC0500009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500168702.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168702.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168702.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168702.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168702.hg.1 |
TC0500009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500168704.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168704.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168704.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168704.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168704.hg.1 |
TC0500009749.hg.1 |
Clariom D TC |