Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004612.hg.1 |
TC16002069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004612.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004612.hg.1 |
ENST00000395384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004612.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004613.hg.1 |
TC16002069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004613.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004613.hg.1 |
ENST00000395384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004613.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004611.hg.1 |
TC16002069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004611.hg.1 |
ENST00000395384.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004611.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600190081.hg.1 |
TC1600011375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600190083.hg.1 |
TC1600011375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000219771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000395384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000562473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
ENST00000564967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011375.hg.1 |
NM_014298.4 |
Refseq |