Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000228.hg.1 |
TC05000009.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000228.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000228.hg.1 |
ENST00000444221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000228.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
ENST00000512038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013144.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192622.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192622.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192622.hg.1 |
ENST00000264935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192622.hg.1 |
NM_018140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192622.hg.1 |
TC0500013144.hg.1 |
Clariom D TC |