Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001454.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001454.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001454.hg.1 |
TC16000074.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600146668.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146668.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146668.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146668.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146668.hg.1 |
TC1600006616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
ENST00000293973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006616.hg.1 |
NM_006181.2 |
Refseq |