Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036797.hg.1 |
TC03002078.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036797.hg.1 |
ENST00000296252.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036797.hg.1 |
ENST00000424591.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036797.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000452897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
ENST00000452897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000429510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000435679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000452897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
ENST00000452897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013417.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
ENST00000296252.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
ENST00000424591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
NM_139248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200009.hg.1 |
TC0300013417.hg.1 |
Clariom D TC |