Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004382.hg.1 |
TC01000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004382.hg.1 |
ENST00000339353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004382.hg.1 |
ENST00000374642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004382.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004382.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004376.hg.1 |
TC01000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004376.hg.1 |
ENST00000339353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004376.hg.1 |
ENST00000374642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004376.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004377.hg.1 |
TC01000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004377.hg.1 |
ENST00000339353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004377.hg.1 |
ENST00000374642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004377.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004386.hg.1 |
TC01000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004386.hg.1 |
ENST00000339353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004386.hg.1 |
ENST00000374642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004386.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004386.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004384.hg.1 |
TC01000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004384.hg.1 |
ENST00000339353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004384.hg.1 |
ENST00000374642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004384.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004384.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004381.hg.1 |
TC01000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004381.hg.1 |
ENST00000339353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004381.hg.1 |
ENST00000374642.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004381.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004381.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150765.hg.1 |
TC0100007290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150766.hg.1 |
TC0100007290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100150760.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150760.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150760.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150760.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150760.hg.1 |
TC0100007290.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000402322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
ENST00000438241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007290.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150759.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150759.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150759.hg.1 |
ENST00000374642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150759.hg.1 |
NM_015991.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150759.hg.1 |
TC0100007290.hg.1 |
Clariom D TC |