Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IWS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017969.2 | NP_060439.2 | 12 - 13 | 2525 | 86 | |
XM_005263707.3 | XP_005263764.1 | 12 - 13 | 2361 | 86 | ||
XM_006712626.3 | XP_006712689.1 | 12 - 13 | 2439 | 86 | ||
XM_017004467.1 | XP_016859956.1 | 12 - 13 | 2526 | 86 | ||
XM_017004468.1 | XP_016859957.1 | 12 - 13 | 2295 | 86 | ||
GenBank mRNA | AK001717.1 | - | 7 - 8 | 770 | 86 | |
AK027561.1 | - | 12 - 13 | 2482 | 86 | ||
AK056881.1 | - | 12 - 13 | 1526 | 86 | ||
AK294669.1 | - | 13 - 14 | 2233 | 86 | ||
AK296848.1 | - | 13 - 14 | 2333 | 86 | ||
AL834178.1 | - | 12 - 13 | 2473 | 86 | ||
BC017012.2 | - | 9 - 10 | 1002 | 86 | ||
BC110536.1 | - | 12 - 13 | 2355 | 40035173 | 86 | |
BC110537.1 | - | 12 - 13 | 2355 | 40035174 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |