Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001041.hg.1 |
ENST00000293805.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001041.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001041.hg.1 |
TC17000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000573503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000573503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000573503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146480.hg.1 |
TC1700006720.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146479.hg.1 |
TC1700006720.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700146481.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146481.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146481.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146481.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146481.hg.1 |
TC1700006720.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000293805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000537931.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000571729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000572216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000573503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000573503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000573503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
ENST00000576705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006720.hg.1 |
NM_181844.3 |
Refseq |