Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003972.2 | NP_003963.1 | 27 - 28 | 4029 | 120 | |
XM_011540326.1 | XP_011538628.1 | 27 - 28 | 4128 | 120 | ||
XM_011540327.2 | XP_011538629.1 | 24 - 25 | 3794 | 120 | ||
XM_011540328.2 | XP_011538630.1 | 24 - 25 | 3622 | 120 | ||
XM_017016877.1 | XP_016872366.1 | 26 - 27 | 4091 | 120 | ||
XM_017016878.1 | XP_016872367.1 | 26 - 27 | 3921 | 120 | ||
XM_017016879.1 | XP_016872368.1 | 26 - 27 | 4045 | 120 | ||
XM_017016880.1 | XP_016872369.1 | 26 - 27 | 4049 | 120 | ||
XM_017016881.1 | XP_016872370.1 | 25 - 26 | 3898 | 120 | ||
GenBank mRNA | AB208843.1 | - | 15 - 16 | 2423 | 120 | |
AF038362.1 | - | 27 - 28 | 4048 | 120 | ||
AJ001017.2 | - | 27 - 28 | 3943 | 120 | ||
AK303554.1 | - | 5 - 6 | 706 | 120 | ||
BC029930.1 | - | 3 - 4 | 424 | 120 | ||
BC112201.1 | - | 27 - 28 | 4013 | 8327669 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |