Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018957.hg.1 |
TC09001514.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018957.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018957.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011294.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
ENST00000436752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
ENST00000612244.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
NM_001199233.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177032.hg.1 |
TC0900011294.hg.1 |
Clariom D TC |