Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014822.hg.1 |
ENST00000357450.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014822.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014822.hg.1 |
TC22000875.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200165104.hg.1 |
TC2200008974.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165104.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165104.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165104.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165104.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000357450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
ENST00000404354.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_001291501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008974.hg.1 |
NM_148674.4 |
Refseq |