Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8033024 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8033023 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1552271_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1552272_PM_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11728629_PM_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11728629_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
TC1900011848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203669.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203668.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203668.hg.1 |
TC1900011848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203668.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
TC1900011848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203667.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
TC1900011848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203671.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900203670.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900203670.hg.1 |
TC1900011848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900203670.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000390672.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
ENST00000419421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011848.hg.1 |
NM_001134316.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1552271_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1552272_a_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1552271_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1552272_PM_a_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552272_a_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1552271_at7 |
- |
- |
Human Genome U95 Set |
71400_PM_at |
- |
- |
Human Genome U95 Set |
71400_at |
- |
- |