Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008758.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008758.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008758.hg.1 |
TC15000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008760.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008760.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008760.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008760.hg.1 |
TC15000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008757.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008757.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008757.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008757.hg.1 |
TC15000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161347.hg.1 |
TC1500008454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161351.hg.1 |
TC1500008454.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000558553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_001304806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008454.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
ENST00000328504.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
ENST00000560327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
ENST00000561002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
NM_173499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161348.hg.1 |
TC1500008454.hg.1 |
Clariom D TC |