Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199865.1 | NP_001186794.1 | 14 - 15 | 1637 | 73 | |
NM_001199866.1 | NP_001186795.1 | 14 - 15 | 1637 | 73 | ||
NM_024704.4 | NP_078980.3 | 14 - 15 | 1637 | 73 | ||
XM_005260750.3 | XP_005260807.1 | 14 - 15 | 1616 | 73 | ||
XM_005260751.4 | XP_005260808.1 | 14 - 15 | 1632 | 73 | ||
XM_005260752.3 | XP_005260809.1 | 14 - 15 | 1616 | 73 | ||
XM_005260753.3 | XP_005260810.1 | 14 - 15 | 1614 | 73 | ||
XM_005260754.3 | XP_005260811.1 | 14 - 15 | 1614 | 73 | ||
XM_005260755.3 | XP_005260812.1 | 14 - 15 | 1660 | 73 | ||
XM_006723588.3 | XP_006723651.1 | 14 - 15 | 1614 | 73 | ||
XM_017027926.1 | XP_016883415.1 | 14 - 15 | 1632 | 73 | ||
GenBank mRNA | AB046810.2 | - | 14 - 15 | 1554 | 73 | |
AK000142.1 | - | 8 - 9 | 879 | 73 | ||
AK095322.1 | - | 14 - 15 | 1574 | 73 | ||
AK307399.1 | - | 14 - 15 | 1520 | 73 | ||
AY166853.1 | - | 14 - 15 | 1636 | 73 | ||
BC050577.1 | - | 14 - 15 | 1637 | 73 | ||
BC150261.1 | - | 14 - 15 | 1554 | 9007204 | 73 | |
BX647572.1 | - | 14 - 15 | 1556 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |