Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003120.hg.1 |
ENST00000374451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003120.hg.1 |
ENST00000458363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003120.hg.1 |
ENST00000476166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003120.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003120.hg.1 |
TC10000272.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007415.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000476166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000479189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000480834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
ENST00000616495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
NM_173160.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000150404.hg.1 |
TC1000007415.hg.1 |
Clariom D TC |