Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005966.hg.1 |
ENST00000278855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005966.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005966.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005966.hg.1 |
TC11000503.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000527395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007762.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
ENST00000278855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
ENST00000532905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
NM_173801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154641.hg.1 |
TC1100007762.hg.1 |
Clariom D TC |