Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006024.hg.1 |
ENST00000270310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006024.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006024.hg.1 |
TC19000460.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
TC1900011710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900200401.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000270310.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000439441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000586063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
ENST00000588265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011710.hg.1 |
NM_022006.1 |
Refseq |