Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021798.hg.1 |
TC07001405.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021798.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021798.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179722.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179722.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179722.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179722.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179722.hg.1 |
TC0700011156.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000388975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
ENST00000477628.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011156.hg.1 |
NM_207366.2 |
Refseq |