Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019326.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019326.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019326.hg.1 |
TC17001459.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019327.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019327.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019327.hg.1 |
TC17001459.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019328.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019328.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019328.hg.1 |
TC17001459.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010604.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183637.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183637.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183637.hg.1 |
ENST00000246672.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700183637.hg.1 |
NM_021724.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700183637.hg.1 |
TC1700010604.hg.1 |
Clariom D TC |