Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026692.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026692.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026692.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026692.hg.1 |
TC11002234.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193249.hg.1 |
TC1100012131.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
ENST00000315274.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_001145938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012131.hg.1 |
NM_002421.3 |
Refseq |