Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278113.1 | NP_001265042.1 | 15 - 16 | 1845 | 134 | |
NM_001464.4 | NP_001455.3 | 16 - 17 | 1902 | 134 | ||
XM_005273468.1 | XP_005273525.1 | 16 - 17 | 1750 | 134 | ||
XM_011544479.1 | XP_011542781.1 | 12 - 13 | 1462 | 134 | ||
XM_011544480.1 | XP_011542782.1 | 12 - 13 | 1372 | 134 | ||
GenBank mRNA | AJ133005.1 | - | 16 - 17 | 1802 | 134 | |
BC034957.2 | - | 15 - 16 | 1765 | 4826530 | 134 | |
BC064547.1 | - | 12 - 13 | 1355 | 5744846 | 134 | |
U38805.1 | - | 16 - 17 | 1866 | 134 | ||
U52370.1 | - | 16 - 17 | 1805 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |