Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018876.hg.1 |
ENST00000308448.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018876.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018876.hg.1 |
ENST00000400215.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018876.hg.1 |
ENST00000541004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018876.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018876.hg.1 |
TC10001492.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018877.hg.1 |
ENST00000308448.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018877.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018877.hg.1 |
ENST00000400215.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018877.hg.1 |
ENST00000541004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018877.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018877.hg.1 |
TC10001492.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000308448.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
ENST00000328142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012575.hg.1 |
NM_032810.3 |
Refseq |