Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007764.hg.1 |
TC03000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007764.hg.1 |
ENST00000288167.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007764.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007762.hg.1 |
TC03000352.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007762.hg.1 |
ENST00000288167.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007762.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154425.hg.1 |
TC0300007523.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007523.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
ENST00000288167.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
ENST00000475124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
ENST00000494338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
NM_018725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154423.hg.1 |
TC0300007523.hg.1 |
Clariom D TC |