Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012348.hg.1 |
ENST00000242592.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012348.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012348.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012348.hg.1 |
TC12000951.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012345.hg.1 |
ENST00000242592.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012345.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012345.hg.1 |
TC12000951.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012347.hg.1 |
ENST00000242592.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012347.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012347.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012347.hg.1 |
TC12000951.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167617.hg.1 |
TC1200009141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167622.hg.1 |
TC1200009141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167619.hg.1 |
TC1200009141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167623.hg.1 |
TC1200009141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009141.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
ENST00000242592.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
ENST00000411593.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
ENST00000539690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
NM_000017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
NM_001302554.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167621.hg.1 |
TC1200009141.hg.1 |
Clariom D TC |