Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020121.3 | NP_064506.3 | 31 - 32 | 3820 | 155 | |
XM_011521094.1 | XP_011519396.1 | 32 - 33 | 3856 | 155 | ||
XM_011521095.1 | XP_011519397.1 | 31 - 32 | 3775 | 155 | ||
XM_011521096.1 | XP_011519398.1 | 31 - 32 | 3724 | 155 | ||
XM_011521097.1 | XP_011519399.1 | 32 - 33 | 3856 | 155 | ||
XM_011521098.1 | XP_011519400.1 | 25 - 26 | 2873 | 155 | ||
XM_011521099.2 | XP_011519401.1 | 24 - 25 | 2790 | 155 | ||
XM_011521102.2 | XP_011519404.1 | 22 - 23 | 2564 | 155 | ||
XM_011521103.2 | XP_011519405.1 | 22 - 23 | 2622 | 155 | ||
XM_017020662.1 | XP_016876151.1 | 30 - 31 | 3643 | 155 | ||
XM_017020663.1 | XP_016876152.1 | 32 - 33 | 3758 | 155 | ||
GenBank mRNA | AF227906.2 | - | 31 - 32 | 3721 | 155 | |
AL133051.1 | - | 28 - 29 | 3200 | 155 | ||
BC125233.1 | - | 31 - 32 | 3676 | 40124676 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |