Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015914.hg.1 |
TC04001160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015914.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015914.hg.1 |
ENST00000540030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04015914.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172174.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172174.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172174.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172174.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172174.hg.1 |
TC0400010596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
ENST00000295452.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010596.hg.1 |
NM_173536.3 |
Refseq |