Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016797.hg.1 |
ENST00000229002.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016797.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016797.hg.1 |
ENST00000538724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016797.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016797.hg.1 |
TC12001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016796.hg.1 |
ENST00000229002.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016796.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016796.hg.1 |
TC12001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016798.hg.1 |
ENST00000229002.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016798.hg.1 |
ENST00000538724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016798.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016798.hg.1 |
TC12001290.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176045.hg.1 |
TC1200010062.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176047.hg.1 |
TC1200010062.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000536890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000538724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000540148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
ENST00000541632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_001286201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010062.hg.1 |
NR_104413.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176044.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176044.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176044.hg.1 |
ENST00000229002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176044.hg.1 |
NM_024730.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176044.hg.1 |
TC1200010062.hg.1 |
Clariom D TC |