Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011527040.2 | XP_011525342.1 | 2 - 3 | 171 | 148 | |
XM_011527041.2 | XP_011525343.1 | 2 - 3 | 171 | 148 | ||
XM_011527042.2 | XP_011525344.1 | 2 - 3 | 171 | 148 | ||
XM_011527043.2 | XP_011525345.2 | 2 - 3 | 171 | 148 | ||
XM_017026884.1 | XP_016882373.1 | 2 - 3 | 172 | 148 | ||
XM_017026885.1 | XP_016882374.1 | 2 - 3 | 175 | 148 | ||
XM_017026886.1 | XP_016882375.1 | 3 - 4 | 1674 | 148 | ||
XM_017026887.1 | XP_016882376.1 | 3 - 4 | 144 | 148 | ||
XM_017026888.1 | XP_016882377.1 | 3 - 4 | 624 | 148 | ||
XM_017026889.1 | XP_016882378.1 | 2 - 3 | 166 | 148 | ||
XM_017026890.1 | XP_016882379.1 | 2 - 3 | 247 | 148 | ||
XM_017026891.1 | XP_016882380.1 | 2 - 3 | 172 | 148 | ||
XM_017026893.1 | XP_016882382.1 | 2 - 3 | 171 | 148 | ||
XM_017026896.1 | XP_016882385.1 | 2 - 3 | 172 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |