Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166448.hg.1 |
TC1700008888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166447.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166447.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166447.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166447.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166447.hg.1 |
TC1700008888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000245543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000579096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000581078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000582136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
ENST00000584947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304272.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_001304273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008888.hg.1 |
NM_024585.3 |
Refseq |