Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014049.hg.1 |
TC01000892.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014049.hg.1 |
ENST00000263174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014049.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014049.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165418.hg.1 |
TC0100009183.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000263174.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000496843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000605613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
ENST00000615664.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009183.hg.1 |
NM_017734.4 |
Refseq |