Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012219.hg.1 |
ENST00000216330.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012219.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012219.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012219.hg.1 |
TC14001084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166600.hg.1 |
TC1400009065.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000216330.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000396062.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000556231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
ENST00000557324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009065.hg.1 |
NM_002013.3 |
Refseq |