Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002946.hg.1 |
ENST00000359332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002946.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002946.hg.1 |
TC14000277.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002944.hg.1 |
ENST00000359332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002944.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002944.hg.1 |
TC14000277.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149170.hg.1 |
TC1400007093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149166.hg.1 |
TC1400007093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149167.hg.1 |
TC1400007093.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000359332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000553274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000554512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000555704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000556392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
ENST00000557128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007093.hg.1 |
NM_172193.2 |
Refseq |