Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000377115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000377115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000377115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000377115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000377115.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000419308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000419308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000419308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000419308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
ENST00000419308.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_021784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_021784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_021784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_021784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_021784.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_153675.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_153675.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_153675.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_153675.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008636.hg.1 |
NM_153675.2 |
Refseq |