Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003128.hg.1 |
TC05000254.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003128.hg.1 |
ENST00000282878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003128.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003129.hg.1 |
TC05000254.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003129.hg.1 |
ENST00000282878.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003129.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000513316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
ENST00000513316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150637.hg.1 |
TC0500007494.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000381158.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000513316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
ENST00000513316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007494.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
ENST00000282878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
ENST00000507977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
NM_138453.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500150634.hg.1 |
TC0500007494.hg.1 |
Clariom D TC |