Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199600.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199600.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199600.hg.1 |
TC1900011677.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199600.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199600.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |