Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011289.hg.1 |
TC05000797.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011289.hg.1 |
ENST00000265271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011289.hg.1 |
ENST00000506502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011289.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000508019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000508019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
ENST00000508019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000508019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000508019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
ENST00000508019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013253.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194488.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194488.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194488.hg.1 |
ENST00000265271.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500194488.hg.1 |
NM_018989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500194488.hg.1 |
TC0500013253.hg.1 |
Clariom D TC |