Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000272198.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000295706.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000367240.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000447715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000486360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000594656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000599966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000600426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
NM_001304331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
NM_001304332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000272198.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000295706.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000367240.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000447715.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000486360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000594656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000599966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
ENST00000600426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
NM_001304331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
NM_001304332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182563.hg.1 |
TC0100011243.hg.1 |
Clariom D TC |