Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027292.hg.1 |
TC06002166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027292.hg.1 |
ENST00000358430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027292.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014350.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
ENST00000358430.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
ENST00000367652.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
NM_153235.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202765.hg.1 |
TC0600014350.hg.1 |
Clariom D TC |