Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025949.hg.1 |
TC06002074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025949.hg.1 |
ENST00000334268.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025949.hg.1 |
ENST00000398161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025949.hg.1 |
ENST00000398178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025949.hg.1 |
NM_001251987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025949.hg.1 |
NM_006073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
ENST00000334268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
NM_001251987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
NM_006073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
ENST00000334268.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
NM_001251987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
NM_006073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190142.hg.1 |
TC0600013062.hg.1 |
Clariom D TC |