Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
TC01002825.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
ENST00000302814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
ENST00000341115.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
ENST00000370588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
ENST00000370589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043952.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210015.hg.1 |
TC0100014769.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000341115.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370587.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000370589.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000474447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000475312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
ENST00000490600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_001253693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014769.hg.1 |
NM_018298.10 |
Refseq |