Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145166.1 | NP_001138638.1 | 9 - 10 | 1105 | 122 | |
NM_001145167.1 | NP_001138639.1 | 10 - 11 | 1155 | 122 | ||
NM_005788.3 | NP_005779.1 | 11 - 12 | 1291 | 122 | ||
XM_011519836.2 | XP_011518138.1 | 8 - 9 | 1268 | 122 | ||
XM_017017081.1 | XP_016872570.1 | 10 - 11 | 1027 | 122 | ||
GenBank mRNA | AF059531.1 | - | 10 - 11 | 1018 | 122 | |
AK125039.1 | - | 3 - 4 | 2768 | 122 | ||
AK129984.1 | - | 10 - 11 | 1209 | 122 | ||
AK298185.1 | - | 10 - 11 | 969 | 122 | ||
AK300591.1 | - | 9 - 10 | 919 | 122 | ||
AK301449.1 | - | 11 - 12 | 1123 | 122 | ||
AK307776.1 | - | 7 - 8 | 750 | 122 | ||
BC019339.1 | - | 10 - 11 | 1127 | 122 | ||
BC037544.1 | - | 11 - 12 | 1079 | 5192246 | 122 | |
BC064831.1 | - | 11 - 12 | 1266 | 6458849 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |