Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017583.5 | NP_060053.2 | 3 - 4 | 1348 | 84 | |
GenBank mRNA | AJ249128.1 | - | 3 - 4 | 1165 | 84 | |
AK021823.1 | - | 3 - 4 | 454 | 84 | ||
AK074242.1 | - | 3 - 4 | 1172 | 84 | ||
BC013166.1 | - | 3 - 4 | 1295 | 3924490 | 84 | |
BC024031.2 | - | 3 - 4 | 1195 | 3532065 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |