Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006688.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006688.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006688.hg.1 |
TC11000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006689.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006689.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006689.hg.1 |
TC11000549.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100155846.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155846.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155846.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155846.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155846.hg.1 |
TC1100007840.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007840.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155842.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155842.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155842.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155842.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155842.hg.1 |
TC1100007840.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100155845.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155845.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155845.hg.1 |
ENST00000244926.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155845.hg.1 |
NM_006551.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155845.hg.1 |
TC1100007840.hg.1 |
Clariom D TC |