Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012514.2 | NP_001012532.1 | 3 - 4 | 547 | 87 | |
NM_001287240.1 | NP_001274169.1 | 4 - 5 | 624 | 87 | ||
NM_001287241.1 | NP_001274170.1 | 5 - 6 | 820 | 87 | ||
NM_030926.5 | NP_112188.1 | 4 - 5 | 688 | 87 | ||
GenBank mRNA | AB003629.1 | - | 4 - 5 | 557 | 87 | |
AF271781.1 | - | 4 - 5 | 673 | 87 | ||
AF272043.1 | - | 4 - 5 | 670 | 87 | ||
AK056321.1 | - | 3 - 4 | 494 | 87 | ||
AK075361.1 | - | 4 - 5 | 653 | 87 | ||
AK090975.1 | - | 4 - 5 | 599 | 87 | ||
AK222697.1 | - | 4 - 5 | 635 | 87 | ||
AK303199.1 | - | 4 - 5 | 642 | 87 | ||
AK307702.1 | - | 3 - 4 | 456 | 87 | ||
AL136603.1 | - | 4 - 5 | 629 | 87 | ||
AL713651.1 | - | 4 - 5 | 612 | 87 | ||
AY049777.1 | - | 4 - 5 | 574 | 87 | ||
BC002424.1 | - | 4 - 5 | 615 | 3162430 | 87 | |
BC050668.1 | - | 3 - 4 | 482 | 5759920 | 87 | |
BC098563.1 | - | 4 - 5 | 601 | 30558635 | 87 | |
CR457208.1 | - | 4 - 5 | 557 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |