Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181712.4 | NP_859063.3 | 3 | 1180 | 135 | |
XM_011540843.2 | XP_011539145.1 | 3 | 1065 | 135 | ||
XM_011540845.2 | XP_011539147.1 | 3 | 1131 | 135 | ||
XM_017000483.1 | XP_016855972.1 | 3 | 1130 | 135 | ||
GenBank mRNA | AB284127.1 | - | 3 | 1180 | 135 | |
BC060866.1 | - | 3 | 1180 | 30344114 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |