Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005549.hg.1 |
TC05000392.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005549.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005549.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005549.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007895.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
ENST00000446378.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
ENST00000505466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
ENST00000506603.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
NM_153610.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500154040.hg.1 |
TC0500007895.hg.1 |
Clariom D TC |