Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008125.hg.1 |
TC10002958.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008125.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008125.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008126.hg.1 |
TC10002958.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008126.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008126.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012490.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186133.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186133.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186133.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186133.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186133.hg.1 |
TC1000012490.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000186134.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186134.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186134.hg.1 |
ENST00000370631.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186134.hg.1 |
NM_018425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186134.hg.1 |
TC1000012490.hg.1 |
Clariom D TC |