Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK293684.1 | - | 1 - 2 | 60 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |